Assay Details
| Gene Symbol: ITPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271930.3 | NP_001258859.1 | 6 - 7 | 509 | 59 | |
| NR_073522.3 | 5 - 6 | 480 | 59 | |||
| GenBank mRNA | BX932147.1 | - | 1 - 2 | 494 | 59 | |
| BX936070.2 | - | 6 - 7 | 509 | 59 | ||
| CR405775.1 | - | 5 - 6 | 489 | 59 | ||
More Information
Set Membership: |
|

ITPA
Genomic Map