Assay Details
| Gene Symbol: TFF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_416743.4 | XP_416743.1 | 1 - 2 | 157 | 63 | |
| GenBank mRNA | CR407288.1 | - | 1 - 2 | 126 | 63 | |
| KC590328.1 | - | 1 - 2 | 162 | 63 | ||
More Information
Set Membership: |
|

TFF3
Genomic Map