Assay Details
| Gene Symbol: MED27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277275.1 | NP_001264204.1 | 2 - 3 | 378 | 77 | |
| XM_025156065.1 | XP_025011833.1 | 2 - 3 | 377 | 77 | ||
| GenBank mRNA | BX930777.2 | - | 1 - 2 | 93 | 77 | |
| BX933293.2 | - | 2 - 3 | 378 | 77 | ||
| CO770311.1 | - | 2 - 3 | 456 | 77 | ||
| CR523351.1 | - | 2 - 3 | 362 | 77 | ||
More Information
Set Membership: |
|

MED27
Genomic Map