Assay Details
| Gene Symbol: CELF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004943647.3 | XP_004943704.3 | 5 - 6 | 607 | 69 | |
| XM_015279061.2 | XP_015134547.2 | 5 - 6 | 607 | 69 | ||
| XM_015279062.2 | XP_015134548.2 | 5 - 6 | 607 | 69 | ||
| XM_015279063.2 | XP_015134549.1 | 5 - 6 | 495 | 69 | ||
| XM_015279064.2 | XP_015134550.1 | 4 - 5 | 438 | 69 | ||
| XM_015279065.2 | XP_015134551.1 | 4 - 5 | 436 | 69 | ||
| XM_015279066.2 | XP_015134552.2 | 5 - 6 | 604 | 69 | ||
| XM_025153988.1 | XP_025009756.1 | 4 - 5 | 350 | 69 | ||
| XM_425051.6 | XP_425051.5 | 5 - 6 | 607 | 69 | ||
| GenBank mRNA | EF520347.1 | - | 5 - 6 | 484 | 69 | |
More Information
Set Membership: |
|

CELF6
Genomic Map