Assay Details
| Gene Symbol: RPL21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233392.4 | XP_001233393.1 | 4 - 5 | 323 | 137 | |
| XM_417127.6 | XP_417127.1 | 4 - 5 | 278 | 137 | ||
| GenBank mRNA | BX930177.1 | - | 3 - 4 | 255 | 137 | |
| BX932372.1 | - | 365 | 137 | |||
| BX933053.1 | - | 3 - 4 | 262 | 137 | ||
| BX933276.1 | - | 4 - 5 | 343 | 137 | ||
More Information
Set Membership: |
|

RPL21
Genomic Map