Assay Details
| Gene Symbol: RPL21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233392.4 | XP_001233393.1 | 5 - 6 | 480 | 115 | |
| XM_417127.6 | XP_417127.1 | 5 - 6 | 435 | 115 | ||
| GenBank mRNA | BX930111.2 | - | 1 - 2 | 872 | 115 | |
| BX930177.1 | - | 4 - 5 | 412 | 115 | ||
| BX932372.1 | - | 522 | 115 | |||
| BX933053.1 | - | 4 - 5 | 419 | 115 | ||
| CR352531.1 | - | 1 - 2 | 1086 | 115 | ||
More Information
Set Membership: |
|

RPL21
Genomic Map