Assay Details
| Gene Symbol: DNAJC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190896.1 | NP_001177825.1 | 2 - 3 | 256 | 77 | |
| XM_015286159.2 | XP_015141645.2 | 2 - 3 | 178 | 77 | ||
| GenBank mRNA | BX930179.1 | - | 2 - 3 | 256 | 77 | |
| BX930378.2 | - | 2 - 3 | 178 | 77 | ||
| CR352549.1 | - | 2 - 3 | 168 | 77 | ||
More Information
Set Membership: |
|

DNAJC24
Genomic Map