Assay Details
| Gene Symbol: CZH18ORF32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_424453.6 | XP_424453.3 | 1 - 2 | 55 | 56 | |
| GenBank mRNA | BX930811.1 | - | 1 - 2 | 36 | 56 | |
| BX930821.1 | - | 1 - 2 | 38 | 56 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_424453.6 transcripts.
Set Membership: |
|

CZH18ORF32
Genomic Map