Assay Details
| Gene Symbol: LOC422609 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015276476.2 | XP_015131962.1 | 4 - 5 | 71 | ||
| XM_015276477.2 | XP_015131963.1 | 6 - 7 | 71 | |||
| XM_015276478.2 | XP_015131964.1 | 5 - 6 | 71 | |||
| XM_025150504.1 | XP_025006272.1 | 6 - 7 | 71 | |||
| XM_025150505.1 | XP_025006273.1 | 4 - 5 | 71 | |||
| XM_420568.5 | XP_420568.2 | 5 - 6 | 71 | |||
| GenBank mRNA | BX932483.1 | - | 3 - 4 | 368 | 71 | |
| BX934425.2 | - | 1 - 2 | 89 | 71 | ||
More Information
Set Membership: |
|

LOC422609
Genomic Map