Assay Details
| Gene Symbol: PCMT1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277402.1 | NP_001264331.1 | 3 - 4 | 342 | 103 | |
| NM_001277404.1 | NP_001264333.1 | 3 - 4 | 342 | 103 | ||
| GenBank mRNA | BX929703.2 | - | 3 - 4 | 327 | 103 | |
| BX935325.1 | - | 3 - 4 | 305 | 103 | ||
| BX950403.1 | - | 3 - 4 | 305 | 103 | ||
| CD215924.1 | - | 3 - 4 | 342 | 103 | ||
More Information
Set Membership: |
|

PCMT1L
Genomic Map