Assay Details
| Gene Symbol: BTG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_418053.6 | XP_418053.3 | 249 | 95 | ||
| GenBank mRNA | BX950447.1 | - | 227 | 95 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BTG2| Gene Symbol: BTG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_418053.6 | XP_418053.3 | 249 | 95 | ||
| GenBank mRNA | BX950447.1 | - | 227 | 95 | ||
Set Membership: |
|