Assay Details
| Gene Symbol: CHMP4C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199475.1 | NP_001186404.1 | 4 - 5 | 740 | 73 | |
| GenBank mRNA | BX929683.1 | - | 1 - 2 | 50 | 73 | |
| BX930325.1 | - | 5 - 6 | 602 | 73 | ||
| BX950548.1 | - | 4 - 5 | 740 | 73 | ||
More Information
Set Membership: |
|

CHMP4C
Genomic Map