Assay Details
| Gene Symbol: PPTC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415161.5 | XP_415161.1 | 4 - 5 | 2899 | 70 | |
| GenBank mRNA | BX950783.1 | - | 4 - 5 | 875 | 70 | |
More Information
Set Membership: |
|

PPTC7
Genomic Map