Assay Details
| Gene Symbol: ZBED1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015277733.2 | XP_015133219.1 | 1 - 2 | 140 | 57 | |
| GenBank mRNA | CR352519.1 | - | 1 - 2 | 79 | 57 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015277733.2 transcripts.
Set Membership: |
|

ZBED1
Genomic Map