Assay Details
| Gene Symbol: RIBC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_416468.6 | XP_416468.3 | 1 - 2 | 747 | 59 | |
| GenBank mRNA | BX930002.2 | - | 1 - 2 | 110 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RIBC1| Gene Symbol: RIBC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_416468.6 | XP_416468.3 | 1 - 2 | 747 | 59 | |
| GenBank mRNA | BX930002.2 | - | 1 - 2 | 110 | 59 | |
Set Membership: |
|