Assay Details
| Gene Symbol: BBOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004941332.3 | XP_004941389.1 | 4 - 5 | 747 | 61 | |
| XM_015286233.2 | XP_015141719.1 | 4 - 5 | 897 | 61 | ||
| XM_424543.5 | XP_424543.2 | 4 - 5 | 772 | 61 | ||
| GenBank mRNA | BX932252.2 | - | 4 - 5 | 810 | 61 | |
| BX936048.1 | - | 4 - 5 | 814 | 61 | ||
| BX950437.1 | - | 4 - 5 | 780 | 61 | ||
More Information
Set Membership: |
|

BBOX1
Genomic Map