Assay Details
| Gene Symbol: ADM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015286460.2 | XP_015141946.1 | 4 | 1041 | 74 | |
| GenBank mRNA | BX933914.2 | - | 1 | 545 | 74 | |
| BX933960.1 | - | 1 | 545 | 74 | ||
| BX950628.1 | - | 4 | 1003 | 74 | ||
More Information
Set Membership: |
|

ADM
Genomic Map