Assay Details
| Gene Symbol: TNNI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004934839.3 | XP_004934896.1 | 120 | 58 | ||
| XM_419242.5 | XP_419242.3 | 161 | 58 | |||
| GenBank mRNA | BX931462.1 | - | 109 | 58 | ||
| BX932175.2 | - | 116 | 58 | |||
| BX934086.1 | - | 116 | 58 | |||
| BX935028.2 | - | 81 | 58 | |||
| BX935107.2 | - | 103 | 58 | |||
| BX935891.1 | - | 100 | 58 | |||
More Information
Set Membership: |
|

TNNI1
Genomic Map