Assay Details
| Gene Symbol: RRP7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277371.1 | NP_001264300.1 | 4 - 5 | 454 | 118 | |
| GenBank mRNA | BU248955.1 | - | 2 - 3 | 207 | 118 | |
| CR387675.1 | - | 4 - 5 | 441 | 118 | ||
More Information
Set Membership: |
|

RRP7A
Genomic Map