Assay Details
| Gene Symbol: PAPOLB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015287288.2 | XP_015142774.1 | 20 - 21 | 2329 | 138 | |
| XM_015287291.2 | XP_015142777.1 | 20 - 21 | 2326 | 138 | ||
| GenBank mRNA | AF061758.1 | - | 16 - 17 | 1695 | 138 | |
More Information
Set Membership: |
|

PAPOLB
Genomic Map