Assay Details
| Gene Symbol: ETV7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004934881.3 | XP_004934938.1 | 2378 | 108 | ||
| XM_004934882.3 | XP_004934939.1 | 2319 | 108 | |||
| XM_004934884.3 | XP_004934941.1 | 1009 | 108 | |||
| XM_025143802.1 | XP_024999570.1 | 1180 | 108 | |||
| XM_025143803.1 | XP_024999571.1 | 2212 | 108 | |||
| XM_025143804.1 | XP_024999572.1 | 2214 | 108 | |||
| XM_025143805.1 | XP_024999573.1 | 2155 | 108 | |||
| XM_025143806.1 | XP_024999574.1 | 759 | 108 | |||
| GenBank mRNA | BX950624.2 | - | 765 | 108 | ||
More Information
Set Membership: |
|

ETV7
Genomic Map