Assay Details
| Gene Symbol: TRABD2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003643049.4 | XP_003643097.2 | 4 - 5 | 1097 | 111 | |
| XM_004949321.3 | XP_004949378.1 | 3 - 4 | 950 | 111 | ||
| XM_004949322.3 | XP_004949379.1 | 4 - 5 | 1097 | 111 | ||
| GenBank mRNA | CR388835.1 | - | 1 - 2 | 120 | 111 | |
More Information
Set Membership: |
|

TRABD2A
Genomic Map