Assay Details
| Gene Symbol: GALNTL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_420520.6 | XP_420520.2 | 11 - 12 | 2689 | 83 | |
| GenBank mRNA | CR389265.1 | - | 7 - 8 | 1023 | 83 | |
More Information
Set Membership: |
|

GALNTL6
Genomic Map