Assay Details
| Gene Symbol: HRASLS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004943333.3 | XP_004943390.1 | 4 | 1412 | 107 | |
| XM_422714.6 | XP_422714.1 | 4 | 1422 | 107 | ||
| GenBank mRNA | CR389674.1 | - | 1 | 851 | 107 | |
More Information
Set Membership: |
|

HRASLS
Genomic Map