Assay Details
| Gene Symbol: RPL12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277679.1 | NP_001264608.1 | 3 - 4 | 262 | 75 | |
| GenBank mRNA | CR390036.1 | - | 3 - 4 | 262 | 75 | |
More Information
Set Membership: |
|

RPL12
Genomic Map