Assay Details
| Gene Symbol: RPL12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277679.1 | NP_001264608.1 | 5 - 6 | 435 | 75 | |
| NR_102416.1 | 5 - 6 | 418 | 75 | |||
| GenBank mRNA | AJ395262.1 | - | 5 - 6 | 418 | 75 | |
| BX934661.2 | - | 4 - 5 | 843 | 75 | ||
| CR390036.1 | - | 5 - 6 | 435 | 75 | ||
More Information
Set Membership: |
|

RPL12
Genomic Map