Assay Details
| Gene Symbol: CHMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_420858.6 | XP_420858.2 | 3 - 4 | 329 | 80 | |
| GenBank mRNA | CR391661.1 | - | 3 - 4 | 272 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHMP3| Gene Symbol: CHMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_420858.6 | XP_420858.2 | 3 - 4 | 329 | 80 | |
| GenBank mRNA | CR391661.1 | - | 3 - 4 | 272 | 80 | |
Set Membership: |
|