Assay Details
| Gene Symbol: PRPS1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_420181.6 | XP_420181.1 | 2 - 3 | 483 | 90 | |
| GenBank mRNA | CR407240.1 | - | 2 - 3 | 477 | 90 | |
More Information
Set Membership: |
|

PRPS1L1
Genomic Map