Assay Details
| Gene Symbol: CUZD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015288891.2 | XP_015144377.2 | 5 - 6 | 1603 | 101 | |
| GenBank mRNA | BX931590.1 | - | 4 - 5 | 663 | 101 | |
| BX931602.2 | - | 5 - 6 | 803 | 101 | ||
More Information
Set Membership: |
|

CUZD1
Genomic Map