Assay Details
| Gene Symbol: LOC421968 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX932813.1 | - | 1 | 746 | 79 | |
| Gene Symbol: TDRD15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015285056.2 | XP_015140542.1 | 3 | 6039 | 79 | |
| XM_025149289.1 | XP_025005057.1 | 3 | 6116 | 79 | ||
More Information
Set Membership: |
|

LOC421968
Genomic Map