Assay Details
| Gene Symbol: KLHL32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015284673.2 | XP_015140159.1 | 9 - 10 | 2740 | 64 | |
| XM_025149236.1 | XP_025005004.1 | 10 - 11 | 1842 | 64 | ||
| GenBank mRNA | BX950519.1 | - | 7 - 8 | 979 | 64 | |
More Information
Set Membership: |
|

KLHL32
Genomic Map