Assay Details
| Gene Symbol: ATP23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199554.1 | NP_001186483.1 | 3 - 4 | 332 | 64 | |
| XM_015281404.2 | XP_015136890.1 | 3 - 4 | 344 | 64 | ||
| XM_025147802.1 | XP_025003570.1 | 2 - 3 | 372 | 64 | ||
| XM_025147804.1 | XP_025003572.1 | 3 - 4 | 405 | 64 | ||
| XM_025147807.1 | XP_025003575.1 | 3 - 4 | 377 | 64 | ||
| XM_025147808.1 | XP_025003576.1 | 3 - 4 | 291 | 64 | ||
| XM_025147815.1 | XP_025003583.1 | 3 - 4 | 392 | 64 | ||
| XM_025147817.1 | XP_025003585.1 | 3 - 4 | 298 | 64 | ||
| XM_025147830.1 | XP_025003598.1 | 3 - 4 | 428 | 64 | ||
| GenBank mRNA | BU356502.1 | - | 3 - 4 | 305 | 64 | |
| BU400702.1 | - | 1 - 2 | 79 | 64 | ||
| BU409185.1 | - | 3 - 4 | 360 | 64 | ||
| CR352492.1 | - | 3 - 4 | 335 | 64 | ||
| CR353901.1 | - | 3 - 4 | 294 | 64 | ||
More Information
Set Membership: |
|

ATP23
Genomic Map