Assay Details
| Gene Symbol: DEPTOR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_417416.5 | XP_417416.2 | 7 - 8 | 1080 | 90 | |
| GenBank mRNA | CR353332.1 | - | 4 - 5 | 614 | 90 | |
More Information
Set Membership: |
|

DEPTOR
Genomic Map