Assay Details
| Gene Symbol: USP41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004937958.3 | XP_004938015.1 | 3 - 4 | 490 | 124 | |
| XM_416398.5 | XP_416398.3 | 3 - 4 | 503 | 124 | ||
| GenBank mRNA | CR354286.1 | - | 3 - 4 | 400 | 124 | |
| LC036549.1 | - | 3 - 4 | 453 | 124 | ||
More Information
Set Membership: |
|

USP41
Genomic Map