Assay Details
| Gene Symbol: VPS33A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025155683.1 | XP_025011451.1 | 5 - 6 | 695 | 74 | |
| XM_415153.5 | XP_415153.1 | 6 - 7 | 799 | 74 | ||
| GenBank mRNA | BX934491.2 | - | 6 - 7 | 764 | 74 | |
More Information
Set Membership: |
|

VPS33A
Genomic Map