Assay Details
| Gene Symbol: TUBAL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277351.1 | NP_001264280.1 | 3 - 4 | 453 | 145 | |
| GenBank mRNA | BU129060.1 | - | 2 - 3 | 404 | 144 | |
| BX935067.2 | - | 3 - 4 | 453 | 145 | ||
More Information
Set Membership: |
|

TUBAL3
Genomic Map