Assay Details
| Gene Symbol: ZNF276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_414213.6 | XP_414213.3 | 4 - 5 | 1082 | 60 | |
| GenBank mRNA | BX936033.2 | - | 1 - 2 | 186 | 60 | |
More Information
Set Membership: |
|

ZNF276
Genomic Map