Assay Details
| Gene Symbol: NMI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025152680.1 | XP_025008448.1 | 7 - 8 | 768 | 72 | |
| GenBank mRNA | BX950337.2 | - | 4 - 5 | 507 | 72 | |
More Information
Set Membership: |
|

NMI
Genomic Map