Assay Details
| Gene Symbol: SSBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278007.1 | NP_001264936.1 | 5 - 6 | 507 | 85 | |
| GenBank mRNA | BU286106.1 | - | 5 - 6 | 458 | 86 | |
| BX932437.1 | - | 5 - 6 | 504 | 85 | ||
| CR352496.1 | - | 5 - 6 | 478 | 85 | ||
More Information
Set Membership: |
|

SSBP1
Genomic Map