Assay Details
| Gene Symbol: SPTSSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302112.1 | NP_001289041.1 | 2 - 3 | 267 | 57 | |
| NM_001302113.1 | NP_001289042.1 | 2 - 3 | 151 | 57 | ||
| XM_015291671.2 | XP_015147157.1 | 2 - 3 | 357 | 57 | ||
| XM_015291672.2 | XP_015147158.1 | 2 - 3 | 284 | 57 | ||
| XM_015291673.2 | XP_015147159.1 | 2 - 3 | 316 | 57 | ||
| XM_015291674.2 | XP_015147160.1 | 2 - 3 | 490 | 57 | ||
| GenBank mRNA | CR353385.1 | - | 2 - 3 | 151 | 57 | |
More Information
Set Membership: |
|

SPTSSB
Genomic Map