Assay Details
| Gene Symbol: C14H16orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015294787.2 | XP_015150273.2 | 23 - 24 | 1962 | 111 | |
| XM_025155367.1 | XP_025011135.1 | 24 - 25 | 2149 | 111 | ||
| XM_025155368.1 | XP_025011136.1 | 24 - 25 | 2061 | 111 | ||
| XM_025155369.1 | XP_025011137.1 | 21 - 22 | 1853 | 111 | ||
| GenBank mRNA | CR385512.1 | - | 1 - 2 | 65 | 111 | |
More Information
Set Membership: |
|

C14H16orf62
Genomic Map