Assay Details
| Gene Symbol: C14H16orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015294787.2 | XP_015150273.2 | 25 - 26 | 2131 | 111 | |
| XM_025155367.1 | XP_025011135.1 | 26 - 27 | 2318 | 111 | ||
| XM_025155368.1 | XP_025011136.1 | 26 - 27 | 2230 | 111 | ||
| XM_025155369.1 | XP_025011137.1 | 23 - 24 | 2022 | 111 | ||
| GenBank mRNA | CR385512.1 | - | 3 - 4 | 234 | 111 | |
More Information
Set Membership: |
|

C14H16orf62
Genomic Map