Assay Details
| Gene Symbol: C14H16orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015294787.2 | XP_015150273.2 | 26 - 27 | 2252 | 74 | |
| XM_025155367.1 | XP_025011135.1 | 27 - 28 | 2439 | 74 | ||
| XM_025155368.1 | XP_025011136.1 | 27 - 28 | 2351 | 74 | ||
| XM_025155369.1 | XP_025011137.1 | 24 - 25 | 2143 | 74 | ||
| GenBank mRNA | CR385512.1 | - | 4 - 5 | 355 | 74 | |
More Information
Set Membership: |
|

C14H16orf62
Genomic Map