Assay Details
| Gene Symbol: EMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277370.2 | NP_001264299.1 | 2 - 3 | 221 | 64 | |
| GenBank mRNA | BM426609.1 | - | 2 - 3 | 221 | 64 | |
| BU307477.1 | - | 2 - 3 | 215 | 64 | ||
| BX930291.2 | - | 2 - 3 | 205 | 64 | ||
| BX930381.2 | - | 2 - 3 | 215 | 64 | ||
More Information
Set Membership: |
|

EMP1
Genomic Map