Assay Details
| Gene Symbol: LYPLAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277567.1 | NP_001264496.1 | 2 - 3 | 216 | 89 | |
| XM_015283652.2 | XP_015139138.1 | 2 - 3 | 538 | 89 | ||
| XM_015283653.2 | XP_015139139.1 | 2 - 3 | 1056 | 89 | ||
| GenBank mRNA | BU372764.1 | - | 2 - 3 | 186 | 89 | |
| BU404580.1 | - | 2 - 3 | 216 | 89 | ||
| BX931356.2 | - | 2 - 3 | 202 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015283653.2 transcripts.
Set Membership: |
|

LYPLAL1
Genomic Map