Assay Details
| Gene Symbol: NSUN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277592.1 | NP_001264521.1 | 4 - 5 | 820 | 103 | |
| GenBank mRNA | BX933119.2 | - | 4 - 5 | 809 | 103 | |
| CR406559.1 | - | 3 - 4 | 408 | 103 | ||
More Information
Set Membership: |
|

NSUN4
Genomic Map