Assay Details
| Gene Symbol: PPP1R14C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015284271.2 | XP_015139757.1 | 4 | 1567 | 142 | |
| XM_015284272.2 | XP_015139758.1 | 5 | 1628 | 142 | ||
| XM_025149191.1 | XP_025004959.1 | 3 | 1542 | 142 | ||
| GenBank mRNA | CR387421.1 | - | 1 | 952 | 142 | |
More Information
Set Membership: |
|

PPP1R14C
Genomic Map