Assay Details
| Gene Symbol: WFIKKN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_420100.5 | XP_420100.2 | 2 | 2213 | 101 | |
| GenBank mRNA | CR388471.1 | - | 1 | 952 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

WFIKKN2| Gene Symbol: WFIKKN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_420100.5 | XP_420100.2 | 2 | 2213 | 101 | |
| GenBank mRNA | CR388471.1 | - | 1 | 952 | 101 | |
Set Membership: |
|