Assay Details
| Gene Symbol: C1QBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025142057.1 | XP_024997825.1 | 5 - 6 | 1370 | 68 | |
| GenBank mRNA | AB029946.1 | - | 4 - 5 | 475 | 68 | |
More Information
Set Membership: |
|

C1QBP
Genomic Map