Assay Details
| Gene Symbol: TBX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270878.1 | NP_001257807.1 | 1 - 2 | 398 | 66 | |
| XM_015294834.2 | XP_015150320.1 | 2 - 3 | 967 | 66 | ||
| GenBank mRNA | AF033669.1 | - | 1 - 2 | 587 | 66 | |
| AF069394.1 | - | 1 - 2 | 59 | 66 | ||
More Information
Set Membership: |
|

TBX3
Genomic Map